Ammonium trifluoromethanesulfonate 98%

Supplier: Ambeed
Warning

A644425-1G A644425-25G
77145-282EA 10.31 USD
77145-282 77145-284
Ammonium trifluoromethanesulfonate 98%
Ammonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: CH₄F₃NO₃S
MDL Number: MFCD00075332
CAS Number: 38542-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR