(1S)-(+)-Camphor-10-sulfonic acid 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid
A644249-25G
A644249-100G
77145-280EA
5.31
USD
77145-280
77617-724
(1S)-(+)-Camphor-10-sulfonic acid 98%
(1S)-(+)-Camphor-10-sulphonic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₆O₄S MW: 232.3 g/mol |
MDL Number:
MFCD00064157 CAS Number: 3144-16-9 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...