3,5-Dibromo-p-toluidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dibromo-4-methylaniline
A643154-1G
A643154-250MG
A643154-5G
77145-272EA
21.63
USD
77145-272
77145-274
77145-276
3,5-Dibromo-p-toluidine 98%
3,5-Dibromo-p-toluidine
Precaución: for Research Use Only
Formula:
C₇H₇Br₂N |
MDL Number:
MFCD00151806 CAS Number: 13194-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...