tert-Butyl(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)dimethylsilane 97%
Supplier: Ambeed
|
Warning
|
A641914-100MG
A641914-1G
A641914-250MG
A641914-5G
77145-254EA
36.08
USD
77145-254
77145-256
77145-258
77145-260
tert-Butyl(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)dimethylsilane 97%
tert-Butyl(4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)dimethylsilane
Precaución: for Research Use Only
Formula:
C₁₈H₃₀BFO₃Si MW: 352.32 g/mol |
MDL Number:
MFCD28515403 CAS Number: 2377611-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...