(2-Iodo-4-nitrophenyl)methanol 97%

Supplier: Ambeed
Warning

A639125-100MG A639125-1G A639125-250MG
77145-244EA 10.3 USD
77145-244 77145-246 77145-248
(2-Iodo-4-nitrophenyl)methanol 97%
(2-Iodo-4-nitrophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆INO₃
MDL Number: MFCD18397985
CAS Number: 500563-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR