3-Borono-4,5-dichlorobenzoic acid 97%

Supplier: Ambeed
Warning

A636070-100MG A636070-1G A636070-250MG
77145-220EA 36.08 USD
77145-220 77145-222 77145-224
3-Borono-4,5-dichlorobenzoic acid 97%
3-Borono-4,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BCl₂O₄
MW: 234.82 g/mol
MDL Number: MFCD29035380
CAS Number: 2377608-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR