(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium 98+%

Supplier: Ambeed
Warning

Synonyms: (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium

A634068-1G A634068-250MG A634068-5G
77145-204EA 46.68 USD
77145-204 77145-206 77145-208
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium 98+%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium

Precaución: for Research Use Only

Formula: C₁₁H₁₉LiO₂
MW: 190.24 g/mol
MDL Number: MFCD00142754
CAS Number: 22441-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR