Methyl 2-hydroxy-5-(trifluoromethyl)benzoate 97%

Supplier: Ambeed
Warning

A633892-100MG A633892-1G A633892-250MG A633892-5G
77145-196EA 45.62 USD
77145-196 77145-198 77145-200 77145-202
Methyl 2-hydroxy-5-(trifluoromethyl)benzoate 97%
Methyl 2-hydroxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
MDL Number: MFCD18399428
CAS Number: 1261478-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR