tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
tert-Butyl (2-chloroethyl)(methyl)carbamate
A633498-1G
A633498-250MG
77145-184EA
98.88
USD
77145-184
77145-186
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate 95%
tert-Butyl N-(2-chloroethyl)-N-methylcarbamate
Precaución: for Research Use Only
Formula:
C₈H₁₆ClNO₂ |
MDL Number:
MFCD20624610 CAS Number: 220074-38-4 UN: 3265 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...