8-Bromo-7-chloro-2-phenyl-[1,2,4]triazolo[1,5-c]pyrimidine 97%

Supplier: Ambeed
Warning

A632615-100MG A632615-1G A632615-250MG A632615-50MG
77145-176EA 112.46 USD
77145-176 77145-178 77145-180 77145-182
8-Bromo-7-chloro-2-phenyl-[1,2,4]triazolo[1,5-c]pyrimidine 97%
8-Bromo-7-chloro-2-phenyl-[1,2,4]triazolo[1,5-c]pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₆BrClN₄
MW: 309.54 g/mol
MDL Number: MFCD18711550
CAS Number: 1254710-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR