Ethyl 6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A632388-100MG A632388-1G A632388-250MG
77145-168EA 95.49 USD
77145-168 77145-170 77145-172
Ethyl 6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate 97%
Ethyl 6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.28 g/mol
MDL Number: MFCD22689252
CAS Number: 1423716-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR