2'-Aminoacetophenone hydrochloride 98%

Supplier: Ambeed
Warning

A631021-10G A631021-1G A631021-250MG A631021-5G
77145-150EA 36.05 USD
77145-150 77145-152 77145-154 77145-156
2'-Aminoacetophenone hydrochloride 98%
2'-Aminoacetophenone hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MDL Number: MFCD00035477
CAS Number: 25384-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR