2-(tert-Butyl)-6-nitro-1H-indole 97%

Supplier: Ambeed
Warning

A626863-100MG A626863-250MG
77145-122EA 50.47 USD
77145-122 77145-124
2-(tert-Butyl)-6-nitro-1H-indole 97%
2-(tert-Butyl)-6-nitro-1H-indole

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
MDL Number: MFCD22689954
CAS Number: 873055-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR