Triflusal 97%

Supplier: Ambeed
Warning

Synonyms: 2-Acetoxy-4-(trifluoromethyl)benzoic acid, Acetyl-4-(trifluoromethyl)salicylic acid, Drisgen

A626746-10MG A626746-1MG A626746-50MG A626746-5MG A626746-250MG
77145-114EA 12.36 USD
77145-114 77145-116 77145-118 77145-120 77686-571
Triflusal 97%
Triflusal

Precaución: for Research Use Only

Formula: C₁₀H₇F₃O₄
MDL Number: MFCD00866793
CAS Number: 322-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR