(3,3,3-Trifluoropropyl)dimethoxymethylsilane 95%

Supplier: Ambeed
Danger

Synonyms: Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane

A626122-10G A626122-25G A626122-5G
77145-100EA 19.1 USD
77145-100 77145-102 77145-104
(3,3,3-Trifluoropropyl)dimethoxymethylsilane 95%
(3,3,3-Trifluoropropyl)dimethoxymethylsilane

Precaución: for Research Use Only

Formula: C₆H₁₃F₃O₂Si
MW: 202.25 g/mol
MDL Number: MFCD00053175
CAS Number: 358-67-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR