(4-Benzyloxycarbonylaminophenyl)boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: Benzyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)carbamate

A624940-1G A624940-250MG A624940-5G
77145-086EA 57.67 USD
77145-086 77145-088 77145-090
(4-Benzyloxycarbonylaminophenyl)boronic acid pinacol ester 96%
(4-Benzyloxycarbonylaminophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₄BNO₄
MW: 353.23 g/mol
MDL Number: MFCD02179440
CAS Number: 363186-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR