1,4-Diamino-2,3-dihydroanthraquinone 99%

Supplier: Ambeed
Warning

Synonyms: 1,4-Diamino-2,3-dihydroanthracene-9,10-dione

A624831-100G A624831-25G A624831-500G A624831-5G
77145-078EA 40.32 USD
77145-078 77145-080 77145-082 77145-084
1,4-Diamino-2,3-dihydroanthraquinone 99%
1,4-Diamino-2,3-dihydroanthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₂
MW: 240.2600 g/mol
MDL Number: MFCD00035689
CAS Number: 81-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR