Dimethyl 2-(3-bromophenyl)malonate 98%

Supplier: Ambeed
Warning

A623361-1G A623361-250MG
77145-070EA 255.68 USD
77145-070 77145-072
Dimethyl 2-(3-bromophenyl)malonate 98%
Dimethyl 2-(3-bromophenyl)malonate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₄
MW: 287.10 g/mol
MDL Number: MFCD06204200
CAS Number: 773134-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR