(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone 97%

Supplier: Ambeed
Warning

Synonyms: (R)-4-Benzhydryloxazolidin-2-one

A623300-100MG A623300-1G
77145-066EA 21.22 USD
77145-066 77145-068
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone 97%
(R)-(+)-4-(Diphenylmethyl)-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO₂
MW: 253.3000 g/mol
MDL Number: MFCD01863565
CAS Number: 173604-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR