Methyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-6-carboxylate 97%

Supplier: Ambeed
Warning

A621146-100MG A621146-250MG
77145-044EA 66.84 USD
77145-044 77145-046
Methyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-6-carboxylate 97%
Methyl 3-bromo-1H-pyrrolo[2,3-b]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.06 g/mol
MDL Number: MFCD21606700
CAS Number: 1638760-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR