1,4-Diazabicyclo[4.3.0]nonane 98%

Supplier: Ambeed
Danger

A619308-100MG A619308-1G A619308-250MG A619308-5G
77145-026EA 25.75 USD
77145-026 77145-028 77145-030 77145-032
1,4-Diazabicyclo[4.3.0]nonane 98%
1,4-Diazabicyclo[4.3.0]nonane

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
MDL Number: MFCD00082600
CAS Number: 5654-83-1
UN: 3267
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR