(4-Nitrobenzyl)triphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: (4-Nitrobenzyl)triphenylphosphonium bromide

A618978-100G A618978-5G
77145-016EA 199.45 USD
77145-016 77145-018
(4-Nitrobenzyl)triphenylphosphonium chloride 98%
(4-Nitrobenzyl)triphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₅H₂₁BrNO₂P
MW: 478.33 g/mol
MDL Number: MFCD00032108
CAS Number: 2767-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR