Z-D-Trp-OBzl 98%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-D-tryptophan Benzyl Ester

A616634-1G A616634-25G A616634-5G
77145-010EA 10.31 USD
77145-010 77145-012 77145-014
Z-D-Trp-OBzl 98%
Z-D-Trp-OBzl

Precaución: for Research Use Only

Formula: C₂₆H₂₄N₂O₄
MDL Number: MFCD01632091
CAS Number: 126496-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR