4-Amino-2,5-difluorobenzonitrile 98%

Supplier: Ambeed
Danger

A616494-1G A616494-25G A616494-5G
77144-996EA 12.36 USD
77144-996 77144-998 77145-000
4-Amino-2,5-difluorobenzonitrile 98%
4-Amino-2,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MDL Number: MFCD00190102
CAS Number: 112279-61-5
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR