2-(Hydroxymethyl)phenylboronic acid cyclic monoester 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydro-1-hydroxy-2,1-benzoxaborole, (2-Hydroxymethylphenyl)boronic acid, dehydrate, 1-Hydroxy-2,1-benzoxaborolane

A616316-1G A616316-250MG A616316-10G A616316-100G
77144-992EA 7.43 USD
77144-992 77144-994 77747-274 77747-273
2-(Hydroxymethyl)phenylboronic acid cyclic monoester 98%
2-(Hydroxymethyl)phenylboronic acid cyclic monoester

Precaución: for Research Use Only

Formula: C₇H₇BO₂
MW: 133.94 g/mol
MDL Number: MFCD01075677
CAS Number: 5735-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR