(6-Ethylpyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A612516-10G A612516-1G A612516-250MG
77144-976EA 1774.68 USD
77144-976 77144-978 77144-980
(6-Ethylpyridin-2-yl)methanol 97%
(6-Ethylpyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD09832885
CAS Number: 163658-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR