Methyl 4-amino-2-cyanobenzoate 97%

Supplier: Ambeed
Warning

A611819-1G A611819-250MG
77144-972EA 117.41 USD
77144-972 77144-974
Methyl 4-amino-2-cyanobenzoate 97%
Methyl 4-amino-2-cyanobenzoate

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MDL Number: MFCD28291983
CAS Number: 1628431-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR