6-(Trifluoromethyl)-1H-indole-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A611221-100MG
A611221-1G
A611221-250MG
77144-960EA
9.27
USD
77144-960
77144-962
77144-964
6-(Trifluoromethyl)-1H-indole-3-carbonitrile 97%
6-(Trifluoromethyl)-1H-indole-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₅F₃N₂ |
MDL Number:
MFCD21607439 CAS Number: 934290-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...