tert-Butyl (6-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A609764-100MG
A609764-1G
A609764-250MG
77144-948EA
35.02
USD
77144-948
77144-950
77144-952
tert-Butyl (6-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate 97%
tert-Butyl (6-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₇H₂₇BN₂O₄ |
MDL Number:
MFCD26520220 CAS Number: 1887223-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...