(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Chloro-3-(hydroxymethyl)phenylboronic acid
A608107-1G
A608107-250MG
A608107-5G
77144-942EA
89.61
USD
77144-942
77144-944
77144-946
(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid 97%
(3-Chloro-5-(hydroxymethyl)phenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₇H₈BClO₃ MW: 186.40 g/mol |
MDL Number:
MFCD22397986 CAS Number: 2121512-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...