5-Bromo-2-methylbenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-methylphenyl)methanol

A607750-1G A607750-250MG A607750-5G
77144-936EA 43.25 USD
77144-936 77144-938 77144-940
5-Bromo-2-methylbenzyl alcohol 95%
5-Bromo-2-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD12031921
CAS Number: 258886-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR