(R)-3-(Naphthalen-1-yloxy)-1-phenylpropan-1-ol 97%
Supplier: Ambeed
|
Warning
|
A606886-100MG
A606886-250MG
77144-930EA
47.38
USD
77144-930
77144-932
(R)-3-(Naphthalen-1-yloxy)-1-phenylpropan-1-ol 97%
(R)-3-(Naphthalen-1-yloxy)-1-phenylpropan-1-ol
Precaución: for Research Use Only
Formula:
C₁₉H₁₈O₂ MW: 278.35 g/mol |
MDL Number:
MFCD22200056 CAS Number: 156453-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...