1-(4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl)ethanone 97%

Supplier: Ambeed
Warning

A606733-100MG A606733-1G A606733-250MG A606733-5G
77144-922EA 36.08 USD
77144-922 77144-924 77144-926 77144-928
1-(4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl)ethanone 97%
1-(4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₂₀H₂₃BO₃
MW: 322.21 g/mol
MDL Number: MFCD22415373
CAS Number: 269410-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR