2-Chloro-5-nitrobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-5-nitrophenyl)methanol

A605619-100G A605619-1G A605619-25G A605619-5G
77144-908EA 258.86 USD
77144-908 77144-910 77144-912 77144-914
2-Chloro-5-nitrobenzyl alcohol 98%
2-Chloro-5-nitrobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD00007297
CAS Number: 80866-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR