3-(Trifluoromethyl)thiophenol 98%

Supplier: Ambeed
Danger

Synonyms: 3-Mercaptobenzotrifluoride, 3-(Trifluoromethyl)benzenethiol, 3-(Trifluoromethyl)benzene-1-thiol

A604793-1G A604793-5G
77144-902EA 18.04 USD
77144-902 77144-904
3-(Trifluoromethyl)thiophenol 98%
3-(Trifluoromethyl)thiophenol

Precaución: for Research Use Only

Formula: C₇H₅F₃S
MW: 178.18 g/mol
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR