4'-Phenoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: p-Phenoxyacetophenone

A601390-100G A601390-10G A601390-25G A601390-5G
77144-880EA 71.09 USD
77144-880 77144-882 77144-884 77144-886
4'-Phenoxyacetophenone 98%
4'-Phenoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
MDL Number: MFCD00008744
CAS Number: 5031-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR