5-Amino-2-mercaptobenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-1H-benzoimidazole-2-thiol

A598733-1G A598733-25G A598733-5G
77144-872EA 6.37 USD
77144-872 77144-874 77144-876
5-Amino-2-mercaptobenzimidazole 98%
5-Amino-2-mercaptobenzimidazole

Precaución: for Research Use Only

Formula: C₇H₇N₃S
MW: 165.22 g/mol
MDL Number: MFCD00022671
CAS Number: 2818-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR