(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-(Diphenylphosphino)phenyl)-4-isopropyl-4,5-dihydrooxazole, (S)-(-)-2-[(2-(Diphenylphosphino)phenyl]-4-(1-methylethyl)-4,5-dihydrooxazole, (S)-iPr-PHOX

A597105-100MG A597105-1G A597105-250MG A597105-5G
77144-866EA 21.22 USD
77144-866 77144-868 77144-870 77551-796
(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole ≥98%, ee 99%
(4S)-(-)-4,5-Dihydro-2-[2'-(diphenylphosphino)phenyl]-4-isopropyloxazole

Precaución: for Research Use Only

Formula: C₂₄H₂₄NOP
MW: 373.436 g/mol
MDL Number: MFCD03427599
CAS Number: 148461-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR