2-Bromo-3-chloro-5-nitropyridine 98%

Supplier: Ambeed
Warning

A595503-1G A595503-25G A595503-5G
77144-852EA 8.24 USD
77144-852 77144-854 77144-856
2-Bromo-3-chloro-5-nitropyridine 98%
2-Bromo-3-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MW: 237.4400 g/mol
MDL Number: MFCD08061302
CAS Number: 22353-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR