(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A594987-100G A594987-10G A594987-25G A594987-500G A594987-5G
77144-836EA 56.65 USD
77144-836 77144-838 77144-840 77144-842 77144-844
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride 98%
(1R,3R)-Methyl 1-(benzo[d][1,3]dioxol-5-yl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₁₉ClN₂O₄
MDL Number: MFCD09842269
CAS Number: 171752-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR