3-Bromo-2-fluoro-6-methylphenylboronic acid 96%

Supplier: Ambeed
Warning

A593134-1G A593134-250MG A593134-5G
77144-818EA 78.28 USD
77144-818 77144-820 77144-822
3-Bromo-2-fluoro-6-methylphenylboronic acid 96%
3-Bromo-2-fluoro-6-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MDL Number: MFCD28515375
CAS Number: 2246650-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR