(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2,2,2-Trifluoro-1-phenylethanol, (S)-(+)-1-Phenyl-2,2,2-trifluoroethanol

A589447-100MG A589447-1G A589447-250MG A589447-25G A589447-5G
77144-800EA 6.18 USD
77144-800 77144-802 77144-804 77144-806 77144-808
(S)-(+)-ɑ-(Trifluoromethyl)benzyl alcohol 98%
(S)-(+)-alpha-(Trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MDL Number: MFCD00077845
CAS Number: 340-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR