(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%

Supplier: Ambeed
Warning

A588047-250MG A588047-5G
77144-796EA 25.76 USD
77144-796 77144-798
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 95%
(R)-(+)-2-Amino-3-methyl-1,1-diphenyl-1-butanol

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO
MDL Number: MFCD01318247
CAS Number: 86695-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR