(±)-3-Fluoro-α-methylbenzylamine 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Fluorophenyl)ethanamine

A586172-1G A586172-250MG
77144-782EA 51.99 USD
77144-782 77144-784
(±)-3-Fluoro-α-methylbenzylamine 95%
(±)-3-Fluoro-α-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD04038308
CAS Number: 74788-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR