Potassium (4-((dimethylamino)methyl)phenyl)trifluoroborate 98%

Supplier: Ambeed
Warning

A583210-1G A583210-250MG A583210-5G
77144-758EA 167.89 USD
77144-758 77144-760 77144-762
Potassium (4-((dimethylamino)methyl)phenyl)trifluoroborate 98%
Potassium (4-((dimethylamino)methyl)phenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₂BF₃KN
MW: 241.1 g/mol
MDL Number: MFCD09993553
CAS Number: 1988699-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR