2-Chloro-4-methylbenzotrifluoride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-methyl-1-(trifluoromethyl)benzene

A582721-100G A582721-10G A582721-1G A582721-25G A582721-5G
77144-736EA 478.47 USD
77144-736 77144-738 77144-740 77144-742 77144-744
2-Chloro-4-methylbenzotrifluoride 97%
2-Chloro-4-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆ClF₃
MW: 194.58 g/mol
MDL Number: MFCD00190112
CAS Number: 74483-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR