1-(tert-Butyl)-1H-1,2,3-triazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A580385-250MG A580385-25G A580385-1G A580385-5G
77144-720EA 12.74 USD
77144-720 77144-722 77686-473 77686-474
1-(tert-Butyl)-1H-1,2,3-triazole-4-carboxylic acid 97%
1-(tert-Butyl)-1H-1,2,3-triazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.18 g/mol
MDL Number: MFCD18250432
CAS Number: 1260665-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR