m-PEG8-acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5,8,11,14,17,20,23-Octaoxahexacosan-26-oic acid, m-PEG7-CH2CH2COOH

A580005-250MG A580005-5G A580005-1G
77144-718EA 49.44 USD
77144-718 77572-846 77594-758
m-PEG8-acid 98%
m-PEG8-acid

Precaución: for Research Use Only

Formula: C₁₈H₃₆O₁₀
MDL Number: MFCD13184957
CAS Number: 1093647-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR