2-Mercaptopyrimidine-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A579375-100MG A579375-1G A579375-250MG
77144-710EA 283.27 USD
77144-710 77144-712 77144-714
2-Mercaptopyrimidine-5-carbaldehyde 95%
2-Mercaptopyrimidine-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₅H₄N₂OS
MW: 140.16 g/mol
MDL Number: MFCD16988189
CAS Number: 856596-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR