(2R,6R)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A579138-100MG A579138-1G A579138-250MG
77144-704EA 46.36 USD
77144-704 77144-706 77144-708
(2R,6R)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid 98%
(2R,6R)-4-(tert-Butoxycarbonyl)-6-methylmorpholine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₅
MDL Number: MFCD30347849
CAS Number: 1581752-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR